Anti-PD1 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: EH12.2H7]

Lieferant: BIOLEGEND INC
329923
BLEG329923EA 370 EUR
BLEG329923
Anti-PD1 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: EH12.2H7]
Antikörper
Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: EH12.2H7
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Common Marmoset, Rhesus, Squirrel Monkey, Cynomolgus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR