Anti-IgD Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: ]

Lieferant: BIOLEGEND INC
348219
BLEG348219EA 304 EUR
BLEG348219
Anti-IgD Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: ]
Antikörper
Type: Primary
Antigen: IGHD
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR