Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (PE (Phycoerythrin))

Lieferant: BIOLEGEND INC
400244
BLEG400244EA 194 EUR
BLEG400244
Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (PE (Phycoerythrin))
Antikörper
Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR