Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (Brilliant Violet® 510)

Lieferant: BIOLEGEND INC
400267
BLEG400267EA 304 EUR
BLEG400267
Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (Brilliant Violet® 510)
Antikörper
Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR