Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (Brilliant Violet® 605)

Lieferant: BIOLEGEND INC
400269
BLEG400269EA 320 EUR
BLEG400269
Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (Brilliant Violet® 605)
Antikörper
Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR