Mouse IgG2b kappa Isotype Control [clone: MPC-11] (Brilliant Violet® 510)

Lieferant: BIOLEGEND INC
400345
BLEG400345EA 304 EUR
BLEG400345
Mouse IgG2b kappa Isotype Control [clone: MPC-11] (Brilliant Violet® 510)
Antikörper
Type: Control
Antigen: IgG2b κ
Clonality: Monoclonal
Clone: MPC-11
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR