Rat IgG1 kappa Isotype Control [clone: RTK2071] (Brilliant Violet® 510)

Lieferant: BIOLEGEND INC
400435
BLEG400435EA 259 EUR
BLEG400435
Rat IgG1 kappa Isotype Control [clone: RTK2071] (Brilliant Violet® 510)
Antikörper
Type: Control
Antigen: IgG1 kappa
Clonality: Monoclonal
Clone: RTK2071
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR