Rat IgG2a kappa Isotype Control [clone: RTK2758] (Brilliant Violet® 650)

Lieferant: BIOLEGEND INC
400541
BLEG400541EA 320 EUR
BLEG400541
Rat IgG2a kappa Isotype Control [clone: RTK2758] (Brilliant Violet® 650)
Antikörper
Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: RTK2758
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: KLH
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR