Rat IgG2b kappa Isotype Control [clone: RTK4530] (PB (Pacific Blue))

Lieferant: BIOLEGEND INC
400627
BLEG400627EA 259 EUR
BLEG400627
Rat IgG2b kappa Isotype Control [clone: RTK4530] (PB (Pacific Blue))
Antikörper
Type: Control
Antigen: IgG2 β
Clonality: Monoclonal
Clone: RTK4530
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR