Mouse IgM kappa Isotype Control [clone: MM-30] (Biotin)

Lieferant: BIOLEGEND INC
401621
BLEG401621EA 102 EUR
BLEG401621
Mouse IgM kappa Isotype Control [clone: MM-30] (Biotin)
Antikörper
Type: Control
Antigen: IgM
Clonality: Monoclonal
Clone: MM-30
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR