Anti-IgG Goat Polyclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
405317
BLEG405317EA 377 EUR
BLEG405317
Anti-IgG Goat Polyclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR