Anti-IgG Goat Polyclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
405414
BLEG405414EA 320 EUR
BLEG405414
Anti-IgG Goat Polyclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol IgG
Antigen-Name Immunoglobulin G
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Brilliant Violet® 421
Reaktivität Mouse
Wirt Goat
Isotyp
ImmunFluoreszenz Yes
Durchflusszytometrie Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR