Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: BIOLEGEND INC
406401
BLEG406401EA 210 EUR
BLEG406401
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR