Anti-IgG Donkey Polyclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
406410
BLEG406410EA 320 EUR
BLEG406410
Anti-IgG Donkey Polyclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Donkey
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR