Anti-IgM Rat Monoclonal Antibody (Biotin)

Lieferant: BIOLEGEND INC
406503
BLEG406503EA 123 EUR
BLEG406503
Anti-IgM Rat Monoclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR