Anti-IgM Rat Monoclonal Antibody [clone: RMM-1] (Brilliant Violet® 421)

Lieferant: BIOLEGEND INC
406532
BLEG406532EA 355 EUR
BLEG406532
Anti-IgM Rat Monoclonal Antibody [clone: RMM-1] (Brilliant Violet® 421)
Antikörper
Bestellen

TECHNISCHE DATEN

Antigen-Symbol IgM
Environmentally Preferable
Antigen-Synonyme Immunoglobulin M
Klonalität Monoklonal
Konjugation Brilliant Violet® 421
Klon RMM-1
Reaktivität Mouse
Wirt Rat
Isotyp IgG2a kappa
Durchflusszytometrie Yes
Immunogen Mouse Ig cocktail
Reinigung The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
Konzentration 0.2 mg/ml
Lagerungspuffer Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Lagertemperatur 2…8 °C
Validierte Anwendungen FC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR