Anti-IGHG1 Rat Monoclonal Antibody (PE (Phycoerythrin)rCP/Cy5.5) [clone: RMG1-1]

Lieferant: BIOLEGEND INC
406612
BLEG406612EA 288 EUR
BLEG406612
Anti-IGHG1 Rat Monoclonal Antibody (PE (Phycoerythrin)rCP/Cy5.5) [clone: RMG1-1]
Antikörper
Type: Primary
Antigen: IGHG1
Clonality: Monoclonal
Clone: RMG1-1
Conjugation: PE (Phycoerythrin)rCP/Cy5.5
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR