Anti-IgG3 Rat Monoclonal Antibody (Biotin)

Lieferant: BIOLEGEND INC
406803
BLEG406803EA 123 EUR
BLEG406803
Anti-IgG3 Rat Monoclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG3
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR