Anti-IgG2 beta Mouse Monoclonal Antibody

Lieferant: BIOLEGEND INC
408202
BLEG408202EA 253 EUR
BLEG408202
Anti-IgG2 beta Mouse Monoclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG2 beta
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR