Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: ]

Lieferant: BIOLEGEND INC
501002
BLEG501002EA 397 EUR
BLEG501002
Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: ]
Antikörper
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR