Anti-IFNG Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 4S.B3]

Lieferant: BIOLEGEND INC
502533
BLEG502533EA 363 EUR
BLEG502533
Anti-IFNG Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 4S.B3]
Antikörper
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 4S.B3
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus
Bestellen

TECHNISCHE DATEN

Antigen-Symbol IFNG
Environmentally Preferable
Antigen-Name Interferon gamma
Antigen-Synonyme AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Brilliant Violet® 570
Klon 4S.B3
Reaktivität Mouse
Wirt Mouse
Isotyp IgG1
Durchflusszytometrie Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR