Anti-IFNG Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 4S.B3]

Lieferant: BIOLEGEND INC
502535
BLEG502535EA 380 EUR
BLEG502535
Anti-IFNG Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 4S.B3]
Antikörper
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 4S.B3
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR