Anti-IL1B Hamster Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
503501 503502
BLEG503501EA 102 EUR
BLEG503501 BLEG503502
Anti-IL1B Hamster Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR