Anti-IL4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
504119
BLEG504119EA 304 EUR
BLEG504119
Anti-IL4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR