Anti-IFNG Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: ]

Lieferant: BIOLEGEND INC
505839
BLEG505839EA 370 EUR
BLEG505839
Anti-IFNG Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: ]
Antikörper
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR