Anti-GZMA Mouse Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
507202
BLEG507202EA 312 EUR
BLEG507202
Anti-GZMA Mouse Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: Granzyme A
Clonality: Monoclonal
Clone:
Conjugation: PURE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR