Anti-IL17A Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: BL168]

Lieferant: BIOLEGEND INC
512325
BLEG512325EA 387 EUR
BLEG512325
Anti-IL17A Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: BL168]
Antikörper
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: BL168
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR