Anti-IL17A Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: BL168]

Lieferant: BIOLEGEND INC
512327
BLEG512327EA 387 EUR
BLEG512327
Anti-IL17A Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: BL168]
Antikörper
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: BL168
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR