Anti-CEBPB Mouse Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
606202
BLEG606202EA 320 EUR
BLEG606202
Anti-CEBPB Mouse Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: C/EBP beta (2 isoforms C/EBP beta)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR