Anti-ANXA5 Antibody (Brilliant Violet® 510) [clone: ]

Lieferant: BIOLEGEND INC
640937
BLEG640937EA 288 EUR
BLEG640937
Anti-ANXA5 Antibody (Brilliant Violet® 510) [clone: ]
Antikörper
Type: Primary
Antigen: ANXA5
Clonality:
Clone:
Conjugation: Brilliant Violet® 510
Epitope:
Host:
Isotype:
Reactivity: Mammals
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR