Anti-TBX21 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 4B10]

Lieferant: BIOLEGEND INC
644815 644816
BLEG644815EA 335 EUR
BLEG644815 BLEG644816
Anti-TBX21 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 4B10]
Antikörper
Type: Primary
Antigen: TBX21
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR