Anti-HIV1 p24 Mouse Monoclonal Antibody (CF405M) [clone: p24/661]
Lieferant: Biotium
BNC050661-500 BNC050661-100
BNC050661-500EA 560 EUR
BNC050661-500 BNC050661-100
Anti-HIV1 p24 Mouse Monoclonal Antibody (CF405M) [clone: p24/661]
Antikörper
Human immunodeficiency virus (HIV) is a retrovirus that causes acquired immune deficiency syndrome (AIDS), a condition in humans in which the immune system begins to fail, leading to life-threatening opportunistic infections. HIV mainly infects vital cells in the human immune system such as helper T cells (specifically CD4 T cells), macrophages and dendritic cells. Two species of HIV infect humans: HIV-1 and HIV-2, with HIV-1 being the more virulent strain. The gag gene of human immunodeficiency virus 1 (HIV-1) encodes a precursor protein known as Pr55Gag. The viral protease PR cleaves thisMore Product Information