Anti-PTPRC Mouse Monoclonal Antibody (CF647) [clone: 190-2F2.5]

Lieferant: Biotium
BNC471081-500 BNC471081-100
BNC471081-500EA 560 EUR
BNC471081-500 BNC471081-100
Anti-PTPRC Mouse Monoclonal Antibody (CF647) [clone: 190-2F2.5]
Antikörper
Recognizes a 180-185 kDa protein, identified as isoform of leukocyte common antigen (CD45RO). This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 190-2F2.5
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR