Anti-CD38 Mouse Monoclonal Antibody (CF488A) [clone: AT2]

Lieferant: Biotium
BNC881086-500 BNC881086-100
BNC881086-500EA 560 EUR
BNC881086-500 BNC881086-100
Anti-CD38 Mouse Monoclonal Antibody (CF488A) [clone: AT2]
Antikörper
This MAb reacts with a 45 kDa glycopeptide, which is a type II membrane glycoprotein with a transmembrane sequence near the NH2terminus. CD38 is a type II transmembrane glycoprotein that is present on early B- and T-cell lineages and activated B- and T-cells but is absent from most mature resting peripheral lymphocytes. CD38 is also found on thymocytes, pre-B cells, germinal center B-cells, mitogen-activated T-cells, monocytes and Ig-secreting plasma cells. CD38 is expressed on CD34 cells. The CD34 CD38- population of hematopoietic stems cells defines the most pluripotent cells (e.g. blast colony forming cells).

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT2
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR