Anti-PAX6 Mouse Monoclonal Antibody (CF594) [clone: PAX6/1166]

Lieferant: Biotium
BNC941166-500 BNC941166-100
BNC941166-500EA 560 EUR
BNC941166-500 BNC941166-100
Anti-PAX6 Mouse Monoclonal Antibody (CF594) [clone: PAX6/1166]
Antikörper
Pax genes contain paired domains with strong homology to genes in Drosophila, which are involved in programming early development. Lesions in the Pax-6 gene account for most cases of aniridia, a congenital malformation of the eye, chiefly characterized by iris hypoplasia, which can cause blindness. Pax-6 is involved in other anterior segment malformations besides aniridia, such as Peters anomaly, a major error in the embryonic development of the eye with corneal clouding with variable iridolenticulocorneal adhesions. The Pax-6 gene encodes a transcriptional regulator that recognizes target genes through its paired-type DNA-binding domain. The paired domain is composed of two distinct DNA-binding subdomains, the amino-terminal subdomain and the carboxy-terminal subdomain, which bind respective consensus DNA sequences. The human Pax-6 gene produces two alternatively spliced isoforms that have the distinct structure of the paired domain.

Type: Primary
Antigen: PAX6
Clonality: Monoclonal
Clone: PAX6/1166
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR