Anti-FUT4 Mouse Monoclonal Antibody (Purified) [clone: Leu-M1]

Lieferant: Biotium
BNUB0277-500 BNUB0277-100
BNUB0277-500EA 511 EUR
BNUB0277-500 BNUB0277-100
Anti-FUT4 Mouse Monoclonal Antibody (Purified) [clone: Leu-M1]
Antikörper
CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: Leu-M1
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR