Anti-PGR Mouse Monoclonal Antibody (Purified) [clone: PR484]

Lieferant: Biotium
BNUB0484-500 BNUB0484-100
BNUB0484-500EA 511 EUR
BNUB0484-500 BNUB0484-100
Anti-PGR Mouse Monoclonal Antibody (Purified) [clone: PR484]
Antikörper
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. It is excellent for immunohistochemical staining of formalin/paraffin tissues.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR484
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR