Anti-MME Mouse Monoclonal Antibody (Purified, BSA-free) [clone: FR4D11]
Lieferant: Biotium
BNUM0248-50
BNUM0248-50EA 417 EUR
BNUM0248-50
Anti-MME Mouse Monoclonal Antibody (Purified, BSA-free) [clone: FR4D11]
Antikörper
Recognizes a 100 kDa glycoprotein, identified as CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA). It is a cell surface enzyme with neutral metalloendopeptidase activity, which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: FR4D11
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human