Anti-ANPEP Mouse Monoclonal Antibody (Purified, BSA-free) [clone: B-F10]
Lieferant: Biotium
BNUM0343-50
BNUM0343-50EA 417 EUR
BNUM0343-50
Anti-ANPEP Mouse Monoclonal Antibody (Purified, BSA-free) [clone: B-F10]
Antikörper
Recognizes an integral membrane glycoprotein of 150 kDa, identified as CD13 (also known as aminopeptidase-N). The CD13 antigen is present on most cells of myeloid origin including granulocytes, monocytes, mast cells, and GM-progenitor cells. It is also expressed by the majority of AML, CML in myeloid blast crisis, and in a smaller fraction of lymphoid leukemias. CD13 is absent from normal lymphocytes, platelets and erythrocytes. CD13 is also present on fibroblasts; endothelial cells, epithelial cells from renal proximal tubules and intestinal brush border, bone marrow stromal cells,More Product Information