Anti-B2M Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 246-E9.E7]
Lieferant: Biotium
BNUM1039-50
BNUM1039-50EA 417 EUR
BNUM1039-50
Anti-B2M Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 246-E9.E7]
Antikörper
Recognizes a protein of 12 kDa, identified as β-microglobulin. β-microglobulin non-covalently associates with the 44 kDa chain to form the HLA Class I antigen complex. Human β-2 microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human β-2 microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of β-2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection.
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 246-E9.E7
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human