Anti-HLA-A Mouse Monoclonal Antibody (Purified, BSA-free) [clone: CATA-1]

Lieferant: Biotium
BNUM1073-50
BNUM1073-50EA 417 EUR
BNUM1073-50
Anti-HLA-A Mouse Monoclonal Antibody (Purified, BSA-free) [clone: CATA-1]
Antikörper
This MAb reacts with cells bearing HLA-A25 or HLA-Aw32 antigens. In addition, a reaction was observed with a cell of phenotype A2, Aw31; B17, Bw49. HLA-A, with HLA-B and HLA-C, belongs to major histocompatibility complex (MHC) class I antigens and expresses constitutively on all nucleated cells. HLA system comprises closely linked genes controlling highly polymorphic proteins involved in the presentation of peptides to the T-cell receptor, inhibition of NK cell cytotoxicity, and rejection of tissue allotransplantation. Specific alleles at HLA loci are associated with diseases. This MAb is specifically applicable for typing peripheral T cells for the antigens HLA-A25 and HLA-Aw32.

Type: Primary
Antigen: HLA-A
Clonality: Monoclonal
Clone: CATA-1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR