Anti-CDKN1B Mouse Monoclonal Antibody (Purified, BSA-free) [clone: SX53G8]
Lieferant: Biotium
BNUM1237-50
BNUM1237-50EA 417 EUR
BNUM1237-50
Anti-CDKN1B Mouse Monoclonal Antibody (Purified, BSA-free) [clone: SX53G8]
Antikörper
This antibody recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.
Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: SX53G8
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Monkey, Mouse, Rat