Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-0007R-A680
BOSSBS-0007R-A680EA 890 EUR
BOSSBS-0007R-A680
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
AM and PAMP are potent hypotensive and vasodilatator agents. Numerous actions have been reported most related to the physiologic control of fluid and electrolyte homeostasis. In the kidney, am is diuretic and natriuretic, and both am and pamp inhibit aldosterone secretion by direct adrenal actions. In pituitary gland, both peptides at physiologically relevant doses inhibit basal ACTH secretion. Both peptides appear to act in brain and pituitary gland to facilitate the loss of plasma volume, actions which complement their hypotensive effects in blood vessels.
Type: Primary
Antigen: Adrenomedullin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 122-140/185
Host: Rabbit
Isotype: IgG
Reactivity: Rat