Anti-GRP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-0011R-A488
BOSSBS-0011R-A488EA 893 EUR
BOSSBS-0011R-A488
Anti-GRP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
GRP stimulates gastrin release as well as other gastrointestinal hormones. Operates as a negative feedback regulating fear and established a causal relationship between GRP-receptor gene expression, long-term potentiation, and amygdala-dependent memory for fear (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Gastrin Releasing Peptide
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat