Anti-GRP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0011R-A680
BOSSBS-0011R-A680EA 890 EUR
BOSSBS-0011R-A680
Anti-GRP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
GRP stimulates gastrin release as well as other gastrointestinal hormones. Operates as a negative feedback regulating fear and established a causal relationship between GRP-receptor gene expression, long-term potentiation, and amygdala-dependent memory for fear (By similarity).

Type: Primary
Antigen: Gastrin Releasing Peptide
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 24-50/148
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR