Anti-GRP Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-0011R-CY3
BOSSBS-0011R-CY3EA 893 EUR
BOSSBS-0011R-CY3
Anti-GRP Rabbit Polyclonal Antibody (Cy3®)
Antikörper
GRP stimulates gastrin release as well as other gastrointestinal hormones. Operates as a negative feedback regulating fear and established a causal relationship between GRP-receptor gene expression, long-term potentiation, and amygdala-dependent memory for fear (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gastrin Releasing Peptide
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR