Anti-TYH Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-0016R-A750
BOSSBS-0016R-A750EA 899 EUR
BOSSBS-0016R-A750
Anti-TYH Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Neuronal Marker Tyrosine hydroxylase is involved in the conversion of tyrosine to dopamine. As the rate-limiting enzyme in the synthesis of catecholamines, tyrosine hydroxylase has a key role in the physiology of adrenergic neurons. Tyrosine hydroxylase is regularly used as a marker for dopaminergic neurons, which is particularly relevant for research into Parkinson's disease.
Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-165/528
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others