Anti-Tyrosine Hydroxylase (TYH) Rabbit Polyclonal Antibody (AbBy Fluor® 594)
Lieferant: Bioss
BS-0016R-BF594
BOSSBS-0016R-BF594EA 852 EUR
BOSSBS-0016R-BF594
Anti-Tyrosine Hydroxylase (TYH) Rabbit Polyclonal Antibody (AbBy Fluor® 594)
Antikörper
Neuronal marker tyrosine hydroxylase is involved in the conversion of tyrosine to dopamine. As the rate-limiting enzyme in the synthesis of catecholamines, tyrosine hydroxylase has a key role in the physiology of adrenergic neurons. Tyrosine hydroxylase is regularly used as a marker for dopaminergic neurons, which is particularly relevant for research into Parkinson's disease.
Type: Polyclonal
Antigen: Tyrosine Hydroxylase (TYH)
Clonality: Polyclonal
Clone:
Conjugation: AbBy Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others