Anti-MTNR1A Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0027R-A680
BOSSBS-0027R-A680EA 899 EUR
BOSSBS-0027R-A680
Anti-MTNR1A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.

Type: Primary
Antigen: Melatonin Receptor 1A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-280/350
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR