Anti-PPP2CA + PPP2CB Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-0029R-A647
BOSSBS-0029R-A647EA 901 EUR
BOSSBS-0029R-A647
Anti-PPP2CA + PPP2CB Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
PP2A is the major phosphatase for microtubule-associated proteins (MAPs). PP2A can modulate the activity of phosphorylase B kinase casein kinase 2, mitogen-stimulated S6 kinase, and MAP-2 kinase. Cooperates with SGOL2 to protect centromeric cohesin from separase-mediated cleavage in oocytes specifically during meiosis I (By similarity). Can dephosphorylate SV4 large T antigen and p53/TP53. Activates RAF1 by dephosphorylating it at 'Ser-259'.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PP2A alpha + beta
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR